siRNA Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019195.2 | NP_062068.1 | 8, 9 | 1018 | ||
| XM_006248264.2 | XP_006248326.1 | 9, 10 | 1711 | |||
| XM_006248265.2 | XP_006248327.1 | 10, 11 | 1756 | |||
| GenBank mRNA | BC085740.1 | - | 8, 9 | 1018 | 7189894 | |
| D87659.1 | - | 8, 9 | 928 | |||
| FQ226437.1 | - | 2, 3 | 130 | |||
| FQ232147.1 | - | 4, 5 | 272 | |||
| FQ232168.1 | - | 3, 4 | 210 | |||
| FQ232318.1 | - | 7, 8 | 641 | |||
| FQ233021.1 | - | 2, 3 | 130 | |||
| FQ233572.1 | - | 3, 4 | 190 | |||

Cd47
Genomic Map