siRNA Details
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019206.2 | NP_062079.2 | 11 | 1942 | ||
XM_008767561.1 | XP_008765783.1 | 11 | 1946 | |||
GenBank mRNA | U33472.1 | - | 6 | 1094 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019206.2 | NP_062079.2 | 11 | 1942 | ||
XM_008767561.1 | XP_008765783.1 | 11 | 1946 | |||
GenBank mRNA | U33472.1 | - | 6 | 1094 |