siRNA Details
Gene Symbol: Men1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019208.3 | NP_062081.1 | 2 | 908 | ||
XM_006230769.2 | XP_006230831.1 | 3 | 626 | |||
XM_006230770.2 | XP_006230832.1 | 3 | 717 | |||
XM_006230771.2 | XP_006230833.1 | 3 | 627 | |||
XM_006230772.2 | XP_006230834.1 | 3 | 685 | |||
GenBank mRNA | AB023400.1 | - | 2 | 975 | ||
AF130369.1 | - | 2 | 883 | |||
AF130370.1 | - | 2 | 528 |
