siRNA Details
| Gene Symbol: Pak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019210.1 | NP_062083.1 | 4 | 470 | ||
| XM_006257333.2 | XP_006257395.1 | 6 | 1743 | |||
| XM_006257336.2 | XP_006257398.1 | 5 | 1632 | |||
| XM_006257338.2 | XP_006257400.1 | 5 | 2035 | |||
| XM_006257339.2 | XP_006257401.1 | 5 | 1697 | |||
| XM_008773404.1 | XP_008771626.1 | 7 | 1808 | |||
| GenBank mRNA | U33314.1 | - | 4 | 470 | ||

Pak3
Genomic Map