siRNA Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019211.2 | NP_062084.2 | 6 | 709 | ||
| GenBank mRNA | AF060819.1 | - | 6 | 571 | ||
| AF081196.1 | - | 6 | 709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgrp1| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019211.2 | NP_062084.2 | 6 | 709 | ||
| GenBank mRNA | AF060819.1 | - | 6 | 571 | ||
| AF081196.1 | - | 6 | 709 | |||