siRNA Details
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019213.2 | NP_062086.1 | 5 | 798 | ||
GenBank mRNA | AB016489.1 | - | 5 | 852 | ||
BC062090.1 | - | 5 | 798 | 5621612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019213.2 | NP_062086.1 | 5 | 798 | ||
GenBank mRNA | AB016489.1 | - | 5 | 852 | ||
BC062090.1 | - | 5 | 798 | 5621612 |