siRNA Details
| Gene Symbol: Sgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193568.1 | NP_001180497.1 | 4, 5 | 489 | ||
| NM_001193569.1 | NP_001180498.1 | 4, 5 | 427 | |||
| NM_019232.3 | NP_062105.2 | 4, 5 | 401 | |||
| XM_006227723.1 | XP_006227785.1 | 6, 7 | 1087 | |||
| XM_008758606.1 | XP_008756828.1 | 4, 5 | 597 | |||
| GenBank mRNA | BC078843.1 | - | 4, 5 | 355 | 7130800 | |
| FM042519.1 | - | 4, 5 | 489 | |||
| FM068981.1 | - | 4, 5 | 407 | |||
| FM123924.1 | - | 4, 5 | 394 | |||
| L01624.1 | - | 4, 5 | 406 | |||

Sgk1
Genomic Map