siRNA Details
| Gene Symbol: Dync1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019234.1 | NP_062107.1 | 3 | 286 | ||
| XM_006236026.2 | XP_006236088.1 | 3 | 464 | |||
| GenBank mRNA | X66845.1 | - | 3 | 286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dync1i1| Gene Symbol: Dync1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019234.1 | NP_062107.1 | 3 | 286 | ||
| XM_006236026.2 | XP_006236088.1 | 3 | 464 | |||
| GenBank mRNA | X66845.1 | - | 3 | 286 | ||