siRNA Details
| Gene Symbol: Ggt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019235.2 | NP_062108.2 | 555 | |||
| GenBank mRNA | BC087018.1 | - | 555 | 7191011 | ||
| U76252.1 | - | 292 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggt5| Gene Symbol: Ggt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019235.2 | NP_062108.2 | 555 | |||
| GenBank mRNA | BC087018.1 | - | 555 | 7191011 | ||
| U76252.1 | - | 292 | ||||