siRNA Details
Gene Symbol: Ggt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019235.2 | NP_062108.2 | 961 | |||
GenBank mRNA | BC087018.1 | - | 961 | 7191011 | ||
U76252.1 | - | 698 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ggt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019235.2 | NP_062108.2 | 961 | |||
GenBank mRNA | BC087018.1 | - | 961 | 7191011 | ||
U76252.1 | - | 698 |