siRNA Details
| Gene Symbol: KLK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012315.1 | NP_036447.1 | 3 | 513 | ||
| GenBank mRNA | AY551001.1 | - | 2 | 356 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLK9| Gene Symbol: KLK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012315.1 | NP_036447.1 | 3 | 513 | ||
| GenBank mRNA | AY551001.1 | - | 2 | 356 | ||