siRNA Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M93271.1 | - | 1988 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bckdk| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M93271.1 | - | 1988 | |||