siRNA Details
| Gene Symbol: P2rx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019256.1 | NP_062129.1 | 1 | 197 | ||
| GenBank mRNA | X95882.1 | - | 1 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2rx7| Gene Symbol: P2rx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019256.1 | NP_062129.1 | 1 | 197 | ||
| GenBank mRNA | X95882.1 | - | 1 | 197 | ||