siRNA Details
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130710.1 | NP_001124182.1 | 2 | 93 | ||
| NM_001139499.1 | NP_001132971.1 | 2 | 332 | |||
| NM_012322.2 | NP_036454.1 | 2 | 156 | |||
| NR_024466.1 | 2 | 903 | ||||
| GenBank mRNA | AF182291.1 | - | 2 | 117 | ||
| AJ238097.1 | - | 2 | 110 | |||
| AK022772.1 | - | 2 | 130 | |||
| AK024217.1 | - | 2 | 139 | |||
| BC005938.1 | - | 2 | 116 | 4132686 | ||
| BF667238.1 | - | 2 | 157 | |||
| DA736378.1 | - | 2 | 323 | |||

LSM5
Genomic Map