siRNA Details
| Gene Symbol: Kcna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019270.3 | NP_062143.3 | 1 | 845 | ||
| GenBank mRNA | X16001.1 | - | 852 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcna3| Gene Symbol: Kcna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019270.3 | NP_062143.3 | 1 | 845 | ||
| GenBank mRNA | X16001.1 | - | 852 | |||