siRNA Details
| Gene Symbol: Sema4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019272.1 | NP_062145.1 | 6 | 723 | ||
| GenBank mRNA | AB002563.1 | - | 6 | 723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sema4f| Gene Symbol: Sema4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019272.1 | NP_062145.1 | 6 | 723 | ||
| GenBank mRNA | AB002563.1 | - | 6 | 723 | ||