siRNA Details
| Gene Symbol: Kcnmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019273.1 | NP_062146.1 | 858 | |||
| XM_006246089.2 | XP_006246151.1 | 2364 | ||||
| XM_008767562.1 | XP_008765784.1 | 3316 | ||||
| XM_008767563.1 | XP_008765785.1 | 3183 | ||||
| GenBank mRNA | AF020712.1 | - | 858 | |||
| FJ154955.1 | - | 547 | ||||
| U40602.1 | - | 545 | ||||

Kcnmb1
Genomic Map