siRNA Details
| Gene Symbol: Exoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019277.2 | NP_062150.2 | 2, 3 | 319 | ||
| XM_008760373.1 | XP_008758595.1 | 2, 3 | 373 | |||
| XM_008760374.1 | XP_008758596.1 | 2, 3 | 2122 | |||
| XM_008760375.1 | XP_008758597.1 | 2, 3 | 386 | |||

Exoc6
Genomic Map