siRNA Details
| Gene Symbol: Resp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019278.1 | NP_062151.1 | 5 | 523 | ||
| GenBank mRNA | BC058147.1 | - | 5 | 519 | 6922403 | |
| L25633.1 | - | 5 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Resp18| Gene Symbol: Resp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019278.1 | NP_062151.1 | 5 | 523 | ||
| GenBank mRNA | BC058147.1 | - | 5 | 519 | 6922403 | |
| L25633.1 | - | 5 | 523 | |||