siRNA Details
| Gene Symbol: Gja5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019280.1 | NP_062153.1 | 2 | 463 | ||
| GenBank mRNA | AF021806.1 | - | 2 | 463 | ||
| M83092.1 | - | 1 | 373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gja5| Gene Symbol: Gja5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019280.1 | NP_062153.1 | 2 | 463 | ||
| GenBank mRNA | AF021806.1 | - | 2 | 463 | ||
| M83092.1 | - | 1 | 373 | |||