siRNA Details
| Gene Symbol: Adcy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019285.2 | NP_062158.2 | 5 | 795 | ||
| GenBank mRNA | BC081813.1 | - | 5 | 795 | 7125413 | |
| FQ223268.1 | - | 5 | 744 | |||
| FQ223451.1 | - | 5 | 744 | |||
| M80633.1 | - | 5 | 761 | |||

Adcy4
Genomic Map