siRNA Details
| Gene Symbol: Arpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019289.2 | NP_062162.1 | 6 | 674 | ||
| GenBank mRNA | AF083269.1 | - | 5 | 587 | ||
| BC062027.1 | - | 6 | 674 | 5622004 | ||
| FQ228791.1 | - | 6 | 645 | |||
| FQ228917.1 | - | 6 | 690 | |||
| FQ228971.1 | - | 6 | 659 | |||
| FQ231755.1 | - | 3 | 372 | |||

Arpc1b
Genomic Map