siRNA Details
| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019299.1 | NP_062172.1 | 5 | 872 | ||
| XM_006247107.2 | XP_006247169.1 | 6 | 1184 | |||
| XM_006247108.2 | XP_006247170.1 | 5 | 1180 | |||
| XM_006247109.2 | XP_006247171.1 | 6 | 1183 | |||
| XM_008768108.1 | XP_008766330.1 | 4 | 1399 | |||
| GenBank mRNA | J03583.1 | - | 5 | 872 | ||

Cltc
Genomic Map