siRNA Details
| Gene Symbol: Dgkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019304.1 | NP_062177.1 | 26 | 2640 | ||
| GenBank mRNA | D16100.1 | - | 26 | 2640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkb| Gene Symbol: Dgkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019304.1 | NP_062177.1 | 26 | 2640 | ||
| GenBank mRNA | D16100.1 | - | 26 | 2640 | ||