siRNA Details
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019305.2 | NP_062178.1 | 1, 2 | 708 | ||
GenBank mRNA | M22427.1 | - | 708 | |||
X07285.1 | - | 1, 2 | 429 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019305.2 | NP_062178.1 | 1, 2 | 708 | ||
GenBank mRNA | M22427.1 | - | 708 | |||
X07285.1 | - | 1, 2 | 429 |