siRNA Details
| Gene Symbol: Itpkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019312.2 | NP_062185.2 | 2 | 1634 | ||
| GenBank mRNA | AJ242781.1 | - | 1183 | |||
| X74227.1 | - | 957 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpkb| Gene Symbol: Itpkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019312.2 | NP_062185.2 | 2 | 1634 | ||
| GenBank mRNA | AJ242781.1 | - | 1183 | |||
| X74227.1 | - | 957 | ||||