siRNA Details
| Gene Symbol: Kcnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019314.1 | NP_062187.1 | 1 | 77 | ||
| XM_006254682.2 | XP_006254744.1 | 1 | 1630 | |||
| XM_006254683.2 | XP_006254745.1 | 2 | 2703 | |||
| GenBank mRNA | JN857942.1 | - | 1 | 77 | ||
| U69882.1 | - | 1 | 77 | |||
| Gene Symbol: LOC103690147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772181.1 | XP_008770403.1 | 1 | 1632 | ||
| XM_008772182.1 | XP_008770404.1 | 2 | 2661 | |||

Kcnn2
Genomic Map