siRNA Details
| Gene Symbol: Mafb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019316.1 | NP_062189.1 | 1 | 1867 | ||
| GenBank mRNA | U56241.1 | - | 1 | 1867 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mafb| Gene Symbol: Mafb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019316.1 | NP_062189.1 | 1 | 1867 | ||
| GenBank mRNA | U56241.1 | - | 1 | 1867 | ||