siRNA Details
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019321.2 | NP_062194.1 | 4 | 435 | ||
GenBank mRNA | U67907.1 | - | 4 | 437 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019321.2 | NP_062194.1 | 4 | 435 | ||
GenBank mRNA | U67907.1 | - | 4 | 437 |