siRNA Details
| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019322.2 | NP_062195.2 | 3 | 474 | ||
| GenBank mRNA | BC061983.1 | - | 3 | 456 | 5598863 | |
| U67910.1 | - | 3 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpsab1| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019322.2 | NP_062195.2 | 3 | 474 | ||
| GenBank mRNA | BC061983.1 | - | 3 | 456 | 5598863 | |
| U67910.1 | - | 3 | 473 | |||