siRNA Details
| Gene Symbol: Nr4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019328.3 | NP_062201.2 | 2 | 254 | ||
| XM_006234201.2 | XP_006234263.1 | 3 | 472 | |||
| GenBank mRNA | DQ092629.1 | - | 2 | 185 | ||
| DQ092631.1 | - | 2 | 199 | |||
| DQ092632.1 | - | 2 | 189 | |||
| FQ222109.1 | - | 3 | 493 | |||
| L08595.1 | - | 2 | 254 | |||
| U01146.1 | - | 2 | 185 | |||
| U72345.1 | - | 3 | 478 | |||

Nr4a2
Genomic Map