siRNA Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019328.3 | NP_062201.2 | 2 | 341 | ||
XM_006234201.2 | XP_006234263.1 | 3 | 559 | |||
XM_008761861.1 | XP_008760083.1 | 2 | 475 | |||
GenBank mRNA | DQ092629.1 | - | 2 | 272 | ||
DQ092630.1 | - | 2 | 108 | |||
DQ092631.1 | - | 2 | 286 | |||
DQ092632.1 | - | 2 | 276 | |||
DQ092633.1 | - | 2 | 108 | |||
DQ092634.1 | - | 2 | 108 | |||
FQ222109.1 | - | 3 | 580 | |||
L08595.1 | - | 2 | 341 | |||
U01146.1 | - | 2 | 272 | |||
U72345.1 | - | 3 | 565 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001139509, NM_013613, S53744, BC137715, AB014889, and others.
Panther Classification:
Gene Ontology Categories:
