siRNA Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019329.2 | NP_062202.1 | 2 | 150 | ||
| XM_006236968.2 | XP_006237030.1 | 3 | 338 | |||
| XM_008763197.1 | XP_008761419.1 | 3 | 338 | |||
| GenBank mRNA | U11031.1 | - | 2 | 151 | ||

Cntn3
Genomic Map