siRNA Details
| Gene Symbol: CFAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012337.2 | NP_036469.2 | 5, 6 | 602 | ||
| GenBank mRNA | AF094758.1 | - | 5, 6 | 635 | ||
| AK225566.1 | - | 5, 6 | 648 | |||
| AK225582.1 | - | 5, 6 | 648 | |||
| AK292249.1 | - | 5, 6 | 651 | |||
| AK308262.1 | - | 5, 6 | 645 | |||
| BC027874.1 | - | 5, 6 | 602 | |||
| BC089391.1 | - | 5, 6 | 797 | 30387473 | ||

CFAP45
Genomic Map