siRNA Details
| Gene Symbol: Eif2ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019335.1 | NP_062208.1 | 3 | 335 | ||
| XM_008764425.1 | XP_008762647.1 | 2 | 559 | |||
| XM_008764426.1 | XP_008762648.1 | 3 | 401 | |||
| GenBank mRNA | L29281.1 | - | 3 | 335 | ||

Eif2ak2
Genomic Map