siRNA Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019344.2 | NP_062217.1 | 6, 7 | 773 | ||
| XM_006250015.2 | XP_006250077.1 | 5, 6 | 644 | |||
| XM_006250016.2 | XP_006250078.1 | 6, 7 | 588 | |||
| XM_006250017.2 | XP_006250079.1 | 6, 7 | 938 | |||
| XM_008769675.1 | XP_008767897.1 | 6, 7 | 939 | |||
| XM_008769676.1 | XP_008767898.1 | 6, 7 | 1104 | |||
| GenBank mRNA | AB006013.1 | - | 5, 6 | 682 | ||
| BC089064.1 | - | 6, 7 | 773 | 7317138 | ||
| U32432.1 | - | 1, 2 | 110 | |||

Rgs8
Genomic Map