siRNA Details
| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019345.3 | NP_062218.3 | 25 | 3268 | ||
| GenBank mRNA | U10097.2 | - | 25 | 3272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc12a3| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019345.3 | NP_062218.3 | 25 | 3268 | ||
| GenBank mRNA | U10097.2 | - | 25 | 3272 | ||