siRNA Details
| Gene Symbol: Slc14a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019346.2 | NP_062219.2 | 9 | 1177 | ||
| XM_008772168.1 | XP_008770390.1 | 10 | 1395 | |||
| XM_008772169.1 | XP_008770391.1 | 9 | 2309 | |||
| XM_008772170.1 | XP_008770392.1 | 9 | 1246 | |||
| GenBank mRNA | FQ211495.1 | - | 9 | 1210 | ||
| U81518.1 | - | 9 | 1147 | |||
| X98399.1 | - | 9 | 1177 | |||

Slc14a1
Genomic Map