siRNA Details
| Gene Symbol: Syt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019350.2 | NP_062223.1 | 3 | 466 | ||
| XM_006228287.2 | XP_006228349.1 | 2 | 156 | |||
| GenBank mRNA | BC092198.1 | - | 3 | 466 | ||
| U26402.1 | - | 3 | 269 | |||
| X84884.1 | - | 2 | 219 | |||

Syt5
Genomic Map