siRNA Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019353.2 | NP_062226.2 | 7, 8 | 851 | ||
| XM_008764607.1 | XP_008762829.1 | 6, 7 | 809 | |||
| XM_008764608.1 | XP_008762830.1 | 7, 8 | 845 | |||
| GenBank mRNA | M31655.1 | - | 3, 4 | 377 | ||
| X17396.1 | - | 7, 8 | 850 | |||

Tpo
Genomic Map