siRNA Details
| Gene Symbol: Pdpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019358.1 | NP_062231.1 | 6 | 1097 | ||
| GenBank mRNA | U07797.1 | - | 6 | 1097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdpn| Gene Symbol: Pdpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019358.1 | NP_062231.1 | 6 | 1097 | ||
| GenBank mRNA | U07797.1 | - | 6 | 1097 | ||