siRNA Details
| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019359.1 | NP_062232.1 | 3 | 461 | ||
| GenBank mRNA | BC062020.1 | - | 3 | 464 | 5623281 | |
| U06755.1 | - | 3 | 461 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnn3| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019359.1 | NP_062232.1 | 3 | 461 | ||
| GenBank mRNA | BC062020.1 | - | 3 | 464 | 5623281 | |
| U06755.1 | - | 3 | 461 | |||