siRNA Details
| Gene Symbol: Arc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019361.1 | NP_062234.1 | 1 | 708 | ||
| XM_008765590.1 | XP_008763812.1 | 1 | 1221 | |||
| XM_008765591.1 | XP_008763813.1 | 1 | 1221 | |||
| GenBank mRNA | U19866.1 | - | 1 | 708 | ||
| Z46925.1 | - | 1 | 689 | |||

Arc
Genomic Map