siRNA Details
| Gene Symbol: Ppt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019367.2 | NP_062240.1 | 9 | 1286 | ||
| XM_006256017.1 | XP_006256079.1 | 9 | 1601 | |||
| XM_006256018.2 | XP_006256080.1 | 9 | 2423 | |||
| XM_006256019.2 | XP_006256081.1 | 9 | 2156 | |||
| XM_006256020.2 | XP_006256082.1 | 9 | 2011 | |||
| XM_006256022.2 | XP_006256084.1 | 9 | 1970 | |||
| XM_006256023.2 | XP_006256085.1 | 9 | 1444 | |||
| XM_008772762.1 | XP_008770984.1 | 9 | 2910 | |||
| GenBank mRNA | AF061971.1 | - | 9 | 1218 | ||
| AF067790.1 | - | 6 | 608 | |||
| BC062023.1 | - | 9 | 1248 | 5623515 | ||
| FQ234132.1 | - | 9 | 1228 | |||

Ppt2
Genomic Map