siRNA Details
| Gene Symbol: Nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019380.1 | NP_062253.1 | 9 | 1913 | ||
| XM_006243172.2 | XP_006243234.1 | 9 | 1936 | |||
| XM_006243174.2 | XP_006243236.1 | 8 | 1597 | |||
| XM_008766330.1 | XP_008764552.1 | 8 | 1585 | |||
| GenBank mRNA | BC070947.1 | - | 8 | 1441 | 7103647 | |
| FQ226464.1 | - | 4 | 796 | |||
| X99337.1 | - | 8 | 1628 | |||
| X99338.1 | - | 9 | 1913 | |||

Nptn
Genomic Map