siRNA Details
| Gene Symbol: Haao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020076.2 | NP_064461.1 | 2, 3 | 255 | ||
| XM_006239684.2 | XP_006239746.1 | 2, 3 | 1160 | |||
| GenBank mRNA | BC085739.1 | - | 2, 3 | 209 | 7108551 | |
| D44494.1 | - | 2, 3 | 256 | |||
| FQ219702.1 | - | 2, 3 | 269 | |||

Haao
Genomic Map