siRNA Details
| Gene Symbol: Rnase4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020082.2 | NP_064467.1 | 2 | 321 | ||
| XM_006251906.2 | XP_006251968.1 | 2 | 360 | |||
| XM_006251907.2 | XP_006251969.1 | 2 | 3938 | |||
| GenBank mRNA | AF041066.1 | - | 2 | 332 | ||
| BC070953.1 | - | 2 | 321 | 7103267 | ||
| BC166436.1 | - | 2 | 904 | 7372999 | ||

Rnase4
Genomic Map