siRNA Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020656.2 | NP_065707.2 | 5 | 470 | ||
| GenBank mRNA | AF264765.2 | - | 5 | 470 | ||
| FQ228521.1 | - | 530 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parva| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020656.2 | NP_065707.2 | 5 | 470 | ||
| GenBank mRNA | AF264765.2 | - | 5 | 470 | ||
| FQ228521.1 | - | 530 | ||||