siRNA Details
| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021583.3 | NP_067594.1 | 3 | 650 | ||
| GenBank mRNA | AB041998.1 | - | 3 | 639 | ||
| AB048730.1 | - | 3 | 652 | |||
| BC088101.1 | - | 3 | 608 | |||
| BC105858.1 | - | 3 | 636 | 7456259 | ||

Ptges
Genomic Map