siRNA Details
| Gene Symbol: Gabra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021841.1 | NP_068613.1 | 9 | 1307 | ||
| XM_006246128.2 | XP_006246190.1 | 9 | 1456 | |||
| XM_008767636.1 | XP_008765858.1 | 7 | 1378 | |||
| GenBank mRNA | FQ213316.1 | - | 2 | 397 | ||

Gabra6
Genomic Map